Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Prolactin Monoclonal Antibody (INN-hPRL-1)
GREENER_CHOICE

Catalog No. MA125051
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA125051 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA125051 Supplier Invitrogen™ Supplier No. MA125051
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This product does require antigen retrieval using heat treatment (1mm EDTA, pH 8.0, is recommended) prior to staining of paraffin sections. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Prolactin (PRL) is a pituitary hormone involved in the stimulation of milk production, salt and water regulation, growth, development and reproduction. PRL exerts its influence on different cell types through a signal transduction pathway which begins with the binding of the hormone to a transmembrane PRL receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Prolactin
Applications ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Radioimmune Assays (RIA)
Classification Monoclonal
Clone INN-hPRL-1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide
Gene PRL
Gene Accession No. P01236
Gene Alias AV290867; decidual prolactin; GHA1; growth hormone A1; Human prolactin; lactogenic prehormone (AA -30 to 199); preprolactin; pre-prolactin; Prl; Prl1a1; PRLB; PRLSD1; Prol; Prolactin; prolactin family 1, subfamily a, member 1; prolactin precursor; prolactin protein; RATPRLSD1; RNPROL
Gene Symbols PRL
Host Species Mouse
Immunogen proprietary immunogen.
Purification Method Ion-exchange Chromatography
Quantity 250 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5617
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.