Learn More
Description
Specifications
Specifications
| Antigen | Prolactin R |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 1D3 |
| Conjugate | Unconjugated |
| Dilution | ELISA |
| Formulation | 50% Glycerol, 0.01M PBS, PH 7.4 |
| Gene Alias | delta 4-delta 7/11 truncated prolactin receptor, delta 4-SF1b truncated prolactin receptor, hPRL receptor, hPRLrI, PRL-R, prolactin receptor, prolactin receptor delta 7/11, secreted prolactin binding protein |
| Host Species | Human |
| Immunogen | Recombinant Human Prolactin R protein [UniProt P16471] |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
