Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Prolactin R Antibody (204), Allophycocyanin

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289377APC
Description
Prolactin R Monoclonal specifically detects Prolactin R in Human samples. It is validated for Flow Cytometry.Specifications
Prolactin R | |
Monoclonal | |
APC | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
204 | |
Flow Cytometry | |
delta 4-delta 7/11 truncated prolactin receptor, delta 4-SF1b truncated prolactin receptor, hPRL receptor, hPRLrI, PRL-R, prolactin receptor, prolactin receptor delta 7/11, secreted prolactin binding protein | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human Prolactin R (NP_000940.1; Met 1-Asp 234). | |
0.1 mL | |
Apoptosis | |
5618 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction