Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Prolactin R Antibody (SPM213), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP234746AF750
Description
Prolactin R Monoclonal specifically detects Prolactin R in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Prolactin R | |
Monoclonal | |
Alexa Fluor 750 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
PRLR | |
Semi-purified human full-length Prolactin R (Uniprot: P16471 ) | |
0.1 mL | |
Apoptosis | |
It recognizes a protein of 70kDa, identified as prolactin receptor. Prolactin is a pituitary hormone involved in the stimulation of milk production, salt and water regulation, growth, development and reproduction. The initial step in its action is the binding to a specific membrane receptor (prolactin receptor), which belongs to the superfamily of class 1 cytokine receptors. The function of the prolactin receptor is mediated, at least in part, by two families of signaling molecules: Janus kinases and signal transducers and activators of transcription. | |
Store at 4°C in the dark. |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
SPM213 | |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
delta 4-delta 7/11 truncated prolactin receptor, delta 4-SF1b truncated prolactin receptor, hPRL receptor, hPRLrI, PRL-R, prolactin receptor, prolactin receptor delta 7/11, secreted prolactin binding protein | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction