Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Prolactin Receptor (ECD) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112169
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112169 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112169 Supplier Invitrogen™ Supplier No. PA5112169
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Prolactin Receptor (ECD) Polyclonal Antibody for Western Blot, IP

Regulator of p53/TP53 stability and function. Inhibits MDM2-mediated degradation of p53/TP53 possibly by cooperating in part with TXNIP (PubMed:16849563, PubMed:23880345). May be involved transcriptional regulation. In vitro has intrinsic transactivation activity enhanced by EP300. May be a transcriptional activator required for the expression of glycolytic genes (PubMed:19919181, PubMed:9928932). Involved in regulation of cell cycle progression. Proposed to disrupt Rb-E2F binding leading to transcriptional activation of E2F proteins (PubMed:19640839). The cell cycle -regulating function may depend on its RUVBL1-mediated association with the R2TP complex (PubMed:26711270). May play a role in regulation of pre-mRNA splicing (PubMed:24722212). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen Prolactin Receptor (ECD)
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.03% ProClin 300; pH 7
Gene ECD
Gene Accession No. O95905
Gene Alias 5730461K03Rik; Ecd; ecdysoneless cell cycle regulator; ecdysoneless homolog; ecdysoneless homolog (Drosophila); GCR2; HSGT1; Human suppressor of GCR two; protein ecdysoneless homolog; protein SGT1; protein SGT1 homolog; SGT1; SGT1 protein; suppressor of GCR2; suppressor of S. cerevisiae gcr2
Gene Symbols ECD
Host Species Rabbit
Immunogen E. coli-derived Human ECD fragment.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11319
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.