Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Prolactin Receptor Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587960
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587960 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587960 Supplier Invitrogen™ Supplier No. PA587960
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QLPPGKPEIF KCRSPNKETF TCWWRPGTDG GLPTNYSLTY HREGETLMHE CPDYITGGPN SCHFGKQYTS MWRTYIMMVN ATNQMGSSFS DELYVDVTYI VQPDPPLELA VEVKQPEDRK PYLWIKWSPP TLIDLKTGWF TLLYEIRLKP EKAAEWEIHF AGQQTEFKIL SLHPGQKYLV QVRCKPDHGY WSAWSPATFI QIPSDFTMND; Positive Samples: 293T, MCF-7, Mouse ovary; Cellular Location: Membrane, Single-pass type I membrane protein.

Prolactin (PRL) is a pituitary hormone involved in the stimulation of milk production, salt and water regulation, growth, development and reproduction. PRL exerts its influence on different cell types through a signal transduction pathway which begins with the binding of the hormone to a transmembrane PRL receptor. Immunoreactive PRL receptor (PRLR), a member of the cytokine receptor family, varies in size (short and long forms) with tissue source and species, from ~40 kDa to 100 kDa. PRLR consists of at least three separate domains: an extracellular region with 5 cysteines which contains the prolactin binding site, a single transmembrane domain and a cytoplasmic region, the length of which appears to influence ligand binding and regulate cellular function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Prolactin Receptor
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.08 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PRLR
Gene Accession No. P05710, P16471, Q08501
Gene Alias AI987712; cytokine receptor; HPRL; hPRL receptor; hPRLrI; Lactogen receptor; MFAB; pPRLR-SF; Pr-1; Pr-3; Prl R; PRLR; PRL-R; Prlr-rs1; Prolactin receptor; prolactin receptor 2 precursor; prolactin receptor related sequence 1; prolactin receptor short isoform; RATPRLR; secreted prolactin binding protein
Gene Symbols PRLR
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 25-234 of human Prolactin Receptor (NP_0009401).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19116, 24684, 5618
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.