Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Prolactin Receptor Recombinant Rabbit Monoclonal Antibody (113)
GREENER_CHOICE

Catalog No. PIMA531132
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531132 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531132 Supplier Invitrogen™ Supplier No. MA531132
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Rat Prolactin Receptor/PRLR.

Prolactin (PRL) is a pituitary hormone involved in the stimulation of milk production, salt and water regulation, growth, development and reproduction. PRL exerts its influence on different cell types through a signal transduction pathway which begins with the binding of the hormone to a transmembrane PRL receptor. Immunoreactive PRL receptor (PRLR), a member of the cytokine receptor family, varies in size (short and long forms) with tissue source and species, from ~40 kDa to 100 kDa. PRLR consists of at least three separate domains: an extracellular region with 5 cysteines which contains the prolactin binding site, a single transmembrane domain and a cytoplasmic region, the length of which appears to influence ligand binding and regulate cellular function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Prolactin Receptor
Applications ELISA
Classification Recombinant Monoclonal
Clone 113
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene PRLR
Gene Accession No. P05710
Gene Alias AI987712; cytokine receptor; HPRL; hPRL receptor; hPRLrI; Lactogen receptor; MFAB; pPRLR-SF; Pr-1; Pr-3; Prl R; PRLR; PRL-R; Prlr-rs1; Prolactin receptor; prolactin receptor 2 precursor; prolactin receptor related sequence 1; prolactin receptor short isoform; RATPRLR; secreted prolactin binding protein
Gene Symbols PRLR
Host Species Rabbit
Immunogen Recombinant Rat Prolactin Receptor/PRLR Protein (Met1-Asp229).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24684
Target Species Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.