Learn More
Pro-Lab Diagnostics™ Prolex™ - Blue Staph Latex Kits
A rapid, sensitive method for distinguishing Staphylococcus aureus that possess coagulase (clumping factor) and/or protein A from other species of staphylococci
$125.10 - $494.00
Catalog Number | Mfr. No. | Quantity | Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Type | Price | Quantity & Availability | ||||
22-286-169
![]() |
Pro-Lab Diagnostics™
PL.09248 |
48 Cards | Agglutination Card |
Pack of 48 for $125.10
|
|
||||
22-286-203
![]() |
Pro-Lab Diagnostics™
PL.080B |
100 Tests | Blue Staph Latex Kit |
Pack of 100 for $260.00
|
|
||||
22-286-171
![]() |
Pro-Lab Diagnostics™
PL.081B |
300 Tests | Blue Staph Latex Kit |
Pack of 300 for $494.00
|
|
||||
Description
- Rapid clear results within 20 seconds
- Uses blue latex particles against a white background
Specifications
S. aureus Coagulase (Clumping Factor), Protein A |