Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pro-Lab Diagnostics™ Prolex™ E. coli Non-O157 Latex Test Reagent Kit
SDP

Catalog No. 23295697 Shop All Pro-Lab Diagnostics Products
Change view
Click to view available options
Format:
Card
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
23-295-697 Card
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-295-697 Supplier Pro-Lab Diagnostics™ Supplier No. PL.1070
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This E. coli Non-O157 Kit is an easy to use, reliable and affordable way to identify the six most common STEC-producing serotypes in North America.

Although E. coli O157 is the most common cause of STEC illness, it has now been recognized that non-O157 strains of E. coli cause severe human disease that is comparable with that caused by E. coli O157. Of these strains, E. coli O26 is the most commonly isolated and can cause hemolytic uremic syndrome (HUS), as several other serotypes can.

The following strains are the next most commonly isolated: E. coli O103, E. coli O111, E. coli O145, E. coli O45 and E. coli O121. These serotypes have also been shown to cause significant human disease. In the United States, the CDC has recommended that all clinical laboratories screen for STEC.

  • Polystyrene beads are sensitized with immunoglobulins against each of the serotype-specific E. coli somatic antigens (E. coli O26, E. coli O103, E. coli O111, E. coli O145, E. coli O121 or E. coli O45)
  • When coated polystyrene particles are mixed with fresh organisms of the corresponding E. coli serotype, bacteria will bind to the antibody, causing particles to visibly agglutinate (positive reaction)
  • Bacteria not of serotypes O26, O103, O111, O145, O121 or O45 will not bind to antibody and will not result in agglutination (negative reaction)

Specifications

Certifications/Compliance FDA, ISO 13485:2016
Content And Storage Store at 2°C to 8°C
Clia Complexity Moderate
Detection Method Latex Agglutination
Format Card
Sample Type Culture
Sensitivity 1
Type Identification Kit
Includes Six Dropper Bottles with E. coli Somatic Antigens, Negative Control, Mixing Cards and Sticks, Agglutination Vials
Analysis Time <30 sec.
Weight (English) 1 lb.
Operating Temperature Range Room Temperature
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.