Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Proliferation Marker Mouse anti-Human, Clone: JC1, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP30776220UG
Description
Proliferation Marker Monoclonal specifically detects Proliferation Marker in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Proliferation Marker | |
Monoclonal | |
0.2 mg/ml | |
Western Blot 1-2 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Immunohistochemistry-Paraffin | |
Mouse | |
Protein A or G purified | |
RUO | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
JC1 | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
Pokeweed-stimulated lymphocytes | |
20 μg | |
Primary | |
Store at 4°C. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction