Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Proline, USP, 98.5-101.5%, Spectrum™ Chemical
SDP

Catalog No. 18605863 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
5 kg
25 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-605-863 100 g Poly Bottle
18-605-862 25 g Poly Bottle
18-605-864 1 kg Poly Bottle
18-605-865 5 kg Poly Pail
18-605-866 25 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-605-863 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. P1434100GM
Add to Cart
Add to Cart

P1434, 147-85-3, C5H9NO2

Spectrum™ Chemical Proline, USP - All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 147-85-3
Molecular Formula C5H9NO2
Molecular Weight (g/mol) 115.13
MDL Number MFCD00064318
InChI Key ONIBWKKTOPOVIA-UHFFFAOYNA-N
IUPAC Name pyrrolidine-2-carboxylic acid
SMILES OC(=O)C1CCCN1

Specifications

CAS Max % 1
Ignition Residue 0.004
Iron (Fe) 30 ppm
Loss on Drying 0.004
Packaging Poly Bottle
Specific Rotation -84.3° to -86.3°
Sulfate 0.0003
Quantity 100 g
Percent Purity 98.5101.5%
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.