Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Molecular Probes™ ProLong™ Live Antifade Reagent, for live cell imaging

Catalog No. p-7244591
Encompass_Preferred
Click to view available options
Quantity:
1 x 1 mL
5 x 1 mL

Prolong Live Antifade Reagent helps prevent the loss of fluorescent signal (photobleaching) during live cell imaging.

ProLong Live Antifade Reagent helps prevent the loss of fluorescent signal due to photobleaching during live cell imaging through use of enzymes from the plasma membrane of E. coli. These enzymes metabolize the elements that can cause photobleaching. When dyes are illuminated, they can degrade which reduces the sample’s fluorescent intensity and leads to the creation of free radical singlet oxygen which degrades neighboring dye molecules. ProLong Live Antifade Reagent metabolizes this element, increasing the signal intensity and duration while keeping the background signal low. ProLong Live reagent has little-to-no effect on cellular viability, proliferation, or other cellular functions in experiments up to 48 hours. ProLong Live reagent provides photobleach protection in live cell experiments for fluorophores such as GFP, RFP, Hoechst 33342, MitoTracker™, LysoTracker™, and CellTracker™dyes. ProLong Live Antifade Reagent can be added to any cell culture media or buffer that is suitable for fluorescent imaging.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 x 1 mL
Product Type Live Antifade Reagent
Content And Storage When stored at -20°C product is stable for at least 6 months, with up to 4 freeze-thaw cycles. When stored at 2–8°C product should be used within 30 days (a precipitant may form at this temperature).
Shipping Condition Approved for shipment at Room Temperature or on Wet Ice
Product Line ProLong™
Reagent Type Cell Imaging Reagent
Volume (Metric) 1 mL
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.