Learn More
PROM2 Mouse anti-Human, Clone: OTI14D1, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF500351

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes a member of the prominin family of pentaspan membrane glycoproteins. The encoded protein localizes to basal epithelial cells and may be involved in the organization of plasma membrane microdomains. Alternative splicing results in multiple transcript variants.Specifications
PROM2 | |
Monoclonal | |
Unconjugated | |
PROM2 | |
PROML2 | |
Mouse | |
Affinity Chromatography | |
RUO | |
150696 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI14D1 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q8N271 | |
PROM2 | |
Full-length protein expressed in 293T cell transfected with human PROM2 expression vector | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.