Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega 5X Green GoTaq™ Flexi Buffer

Catalog No. PRM8911 Shop All Promega Corporation Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRM8911 20 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PRM8911 Supplier Promega Supplier No. M8911
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

5X Green GoTaq Flexi Reaction Buffers are Mg-free buffers that allow optimization of Mg concentration in PCR. The green buffer allows direct-to-gel analysis of reaction products without the need to add loading dye.

  • Mg-Free Buffers for PCR Optimization
  • Use with GoTaq(R) Flexi DNA Polymerase and Mg Solution to optimize magnesium concentration in PCR
  • Available with green dye for direct-to-gel convenience, or in colorless format for applications where absorbance or fluorescence measurements are necessary prior to PCR cleanup

Specifications

Concentration 5X
Content And Storage Two dyes (a blue dye and a yellow dye) that separate during electrophoresis to show migration progress. Store at −30 to −10°C
pH 8.5
Quantity 20 mL
Volume 4 x 5mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.