Learn More
Promega Amino Acid Mixtures
Catalog No. PRL4461 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Product Type:
Amino Acid Mixture, Complete
Amino Acid Mixture, no Methionine
Amino Acid Mixture, no Methionine or Cysteine
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
PR-L4461 Amino Acid Mixture, Complete
PR-L9961 Amino Acid Mixture, no Methionine
PR-L5511 Amino Acid Mixture, no Methionine or Cysteine
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PR-L4461 Supplier Promega Supplier No. L4461
Add to Cart
Add to Cart
Amino Acid Mixture, Complete, is an aqueous solution containing 1mM each of the 20 essential amino acids.
  • 1mM Solution of 20 Essential Amino Acids or Combinations Thereof
  • Amino acid solution for use with Flexi(R), TnT(R) and Rabbit Reticulocyte Lysates, Wheat Germ Extract and E. coli S30 Systems
  • Choose from mix of 20 amino acids or special mixtures indicated
  • Supplied at a concentration 1mM
Specifications
Content And Storage <−65°C
Product Type Amino Acid Mixture, Complete
Quantity 175 μL