Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Promega AMV Reverse Transcriptase
AMV RT catalyzes DNA polymerization using template DNA, RNA or RNA:DNA hybrids.
Supplier: Promega M5108
Includes: 5X Reaction Buffer (250mM Tris-HCl [pH 8.3 at 25°C], 250mM KCl, 50mM MgCl2, 2.5mM spermidine, 50mM DTT)
Description
- Efficient Reverse Transcription of Templates with High Secondary Structure
- DNA polymerization using template DNA, RNA or RNA:DNA hybrids
- Requires a primer as well as magnesium or manganese
- Stable at higher reaction temperatures (37-58°C)
- Supplied at a concentration of 10u/μL (Cat.# M5101, M5108) or 20-25u/μL (Cat.# M9004)
Specifications
10 U/μL | |
AMV Reverse Transcriptase | |
1000U | |
3 x 500μL 5X reaction buffer | |
Potassium Phosphate (pH 7.2 at °C) 200mM, 0.2% Triton™ X-100, DTT 2mM and Glycerol 50% |
−30 to −10°C | |
Liquid | |
cDNA Synthesis | |
Avian Myeloblastosis Virus |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction