Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Beta-Glo™ Assay System

Catalog No. PRE4740 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 mL
100 mL
10 x 100 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRE4740 100 mL
PRE4720 10 mL
PRE4780 10 x 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PRE4740 Supplier Promega Supplier No. E4740
Add to Cart
Add to Cart

Includes

Beta-Glo Assay Buffer; Beta-Glo Assay Substrate

The Beta-Glo™ Assay System is a homogeneous, luminescence-based method for quantitating beta-galactosidase expression in mammalian cells.

  • Luminescence Detection of β-Gal Activity
  • Add a single reagent directly to cells in growth medium
  • Optimal assay performance at room temperature
  • Read results using any luminometer. Injectors not required

Specifications

Includes 1 x 20mL Beta-Glo™ assay substrate, 1 x 100mL Beta-Glo™ Assay Buffer
pH 7.5
Content And Storage -30°C to -10°C
Quantity 100 mL
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.