Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega BTK Kinase Enzyme System

Catalog No. PRV2941 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1/Ea.
10 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRV2941 10 μg
PRV9071 1/Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PRV2941 Supplier Promega Supplier No. V2941
Add to Cart
Add to Cart
This item is not returnable. View return policy

Includes

  • 1 x 10μg BTK Kinase Enzyme System (V2941) and 1 x 1000 ADP-Glo™ Kinase Assays (V9101)

Recombinant full-length human BTK was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag. BTK (also known as Bruton tyrosine kinase) plays a crucial role in B-lymphocyte differentiation and activation.

  • Easily Screen and Profile BTK Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity

Specifications

Content And Storage <-65°C
For Use With (Application) Both primary screening as well as Kinase selectivity profiling
Quantity 10 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.