Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega c-MER Kinase Enzyme System

Catalog No. PRV3541 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1/Ea.
10 μg
For Use With (Application):
Involved in engulfment and clearance of Apoptotic Cells
Involved in engulfment and clearance of Apoptotic Cells, used to monitor the activity of virtually any ADP-generating Enzyme
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity For Use With (Application)
PRV3541 10 μg Involved in engulfment and clearance of Apoptotic Cells
PRV9561 1/Ea. Involved in engulfment and clearance of Apoptotic Cells, used to monitor the activity of virtually any ADP-generating Enzyme
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRV3541 Supplier Promega Supplier No. V3541
Add to Cart
Add to Cart

Recombinant human c-MER (amino acids 578-872) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. c-MER is a tyrosine kinase and a member of the MER/AXL/TYRO3 receptor kinase family.

  • Easily Screen and Profile c-MER Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity

Specifications

Product Type c-MER Kinase Enzyme System
Content And Storage <-65°C
For Use With (Application) Involved in engulfment and clearance of Apoptotic Cells
Quantity 10 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.