Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega CAMK2gamma Kinase Enzyme System

Catalog No. PRV3531 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1/Ea.
10 μg
Product Type:
ADP-Glo Kinase Assay and CAMK2 gamma Kinase Enzyme System
CAMK2 gamma Kinase Enzyme System
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type
PRV3531 10 μg CAMK2 gamma Kinase Enzyme System
PRV9201 1/Ea. ADP-Glo Kinase Assay and CAMK2 gamma Kinase Enzyme System
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRV3531 Supplier Promega Supplier No. V3531
Add to Cart
Add to Cart

Recombinant human CAMK2 gamma (C-terminal truncation) was expressed by baculovirus in Sf9 cells using an N-terminal GST tag. CAMK2 gamma is a member of the CAMKII family.

  • Easily Screen and Profile CAMK2γ Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity

Specifications

Product Type CAMK2 gamma Kinase Enzyme System
Molecular Weight (g/mol) 60
Content And Storage <-65°C
For Use With (Application) Implicated in diverse effects of hormones and neurotransmitters
Quantity 10 μg
Form Fluid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.