Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega cAMP, 1mM

Catalog No. PRV6421 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PR-V6421 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PR-V6421 Supplier Promega Supplier No. V6421
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

cAMP (adenosine-3',5'-cyclic monophosphate) is an activator of cAMP-Dependent Protein Kinase, Catalytic Subunit.

  • Adenosine-3′,5′-Cyclic Monophosphate
  • Activator of cAMP-Dependent Protein Kinase, catalytic subunit
  • Supplied at 1mM concentration

Specifications

Concentration 1mM
Formulation 1 x 500μL cAMP
For Use With (Application) For laborato ry use
Sample Volume 500μL
Content And Storage −30 to −10°C
Product Type cAMP
Quantity 500 μL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.