Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Caspase-Glo™ 8 and Caspase-Glo™ 9 Assay Systems

Catalog No. PRG8211 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
2.5 mL
10 mL
100 mL
Product Type:
Caspase-Glo 8 Assay
Caspase-Glo 9 Assay
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type
PRG8211 10 mL Caspase-Glo 9 Assay
PRG8201 10 mL Caspase-Glo 8 Assay
PRG8200 2.5 mL Caspase-Glo 8 Assay
PRG8202 100 mL Caspase-Glo 8 Assay
PRG8210 2.5 mL Caspase-Glo 9 Assay
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. PRG8211 Supplier Promega Supplier No. G8211
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Caspase-Glo 8 Buffer or Caspase-Glo 9 Buffer; Caspase-Glo 8 Substrate or Caspase-Glo 9 Substrate

Caspase-Glo™ Assay is a homogeneous luminescent assay that measures caspase-8 or caspase-9 activity.

  • A Luminescent Assay That Measures Caspase-8 or 9 Activity
  • Decreased assay time and less enzyme needed
  • Homogeneous add-mix-measure protocol amenable to automation

  • U.S. Patent 6,602,677, Australian Patent 754312 and other patents pending.
  • The method of recombinant expression of Coleoptera luciferase is covered by U.S. Patents 5,583,024; 5,674,713; and 5,700,673.

Specifications

Quantity 10 mL
Test Time 0.5 to 1 hr.
Product Type Caspase-Glo 9 Assay
Content And Storage 1 x 1 Bottle Caspase-Glo 9 substrate, 1 x 10mL Caspase-Glo 9 Buffer, 1 x 30μL MG-132 inhibitor. Store at -30°C to -10°C
For Use With (Application) Caspase-9 Activation Determination (EC50), Caspase-9 Activity Determination, Caspase-9 Inhibition Determination (IC50)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.