Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega CellTiter-Blue™ Cell Viability Assays

Catalog No. PRG8082 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
20 mL
100 mL
10 x 100 mL
No. of Reactions:
1000 in 96-well Plates or 4000 in 384-well Plates
50,000 in 96-well Plates or 200,000 in 384-well Plates
5000 in 96-well Plates or 20,000 384-well Plates
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity No. of Reactions
PR-G8082 10 x 100 mL 50,000 in 96-well Plates or 200,000 in 384-well Plates
PR-G8081 100 mL 5000 in 96-well Plates or 20,000 384-well Plates
PR-G8080 20 mL 1000 in 96-well Plates or 4000 in 384-well Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PR-G8082 Supplier Promega Supplier No. G8082
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Homogeneous, fluorometric cell viability assay (resazurin). Requires a 1- to 4-hour incubation.

  • A Homogeneous, Fluorescent Assay to Monitor Cell Viability
  • Fast, flexible, time-saving
  • Amenable to multiplexing with other assays

Specifications

Quantity 10 x 100 mL
Format 96- or 384-well Formats
Content And Storage -30°C to -10°C
No. of Reactions 50,000 in 96-well Plates or 200,000 in 384-well Plates
Detection Method Fluorescence (preferred) or Absorbance
For Use With (Application) Monitoring Cell Viability
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.