Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega cGMP, 1mM

Catalog No. PRV6411 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRV6411 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRV6411 Supplier Promega Supplier No. V6411
May include imposed supplier surcharges.
Add to Cart
Add to Cart

cGMP (guanosine-3',5'-cyclic monophosphate) is an activator of cGMP-Dependent Protein Kinase.

  • Guanosine-3′,5′-Cyclic Monophosphate
  • Activator of cGMP-Dependent Protein Kinase
  • Supplied at 1mM concentration

Specifications

Concentration 1mM
For Use With (Application) For laborato ry use
Content And Storage −30 to −10°C
Product Type cGMP
Quantity 500 μL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.