Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Control Vectors for CheckMate™/Flexi™ Vector Mammalian Two-Hybrid Systems

Catalog No. PRC9370 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Vector:
pBIND, pACT neg ctrl
pBIND-Id, pACT-MyoD,pos ctrl
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Vector
PRC9370 pBIND-Id, pACT-MyoD,pos ctrl
PRC9380 pBIND, pACT neg ctrl
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRC9370 Supplier Promega Supplier No. C9370
Add to Cart
Add to Cart

Powerful system for detecting protein:protein interactions in vivo.

  • Powerful System for Detecting Protein:Protein Interactions In Vivo
  • Mammalian system for studying interactions in cell line of choice
  • Study post-translational modifications including glycosylation, phosphorylation and acylation

Specifications

For Use With (Equipment) CheckMate™/Flexi™ vector mammalian two-hybrid system
Storage Buffer 10mM Tris HCl, 1mM EDTA (pH 7.4)
Content And Storage -30°C to -10°C
Concentration 1 μg/μL
Quantity 1 Stab/ 20μg
Vector pBIND-Id, pACT-MyoD,pos ctrl
For Use With (Application) Transcription regulation, virus-cell interactions, compound screening, post-translational modifications, GPCR signaling, cell signaling, promoter analysis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.