Learn More
Promega CSK Kinase Enzyme System
Catalog No. PRV2981 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1/Ea.
10 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRV2981 10 μg
PRV9251 1/Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRV2981 Supplier Promega Supplier No. V2981
Only null left
Add to Cart
Add to Cart
Includes
  • 1 x 10μg CSK Kinase Enzyme System (V2981) and 1 x 1000 ADP-Glo™ Kinase Assays (V9101)
Recombinant full-length human CSK was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. CSK has been shown to downregulate the tyrosine kinase activity of c-src through tyrosine phosphorylation of the c-src carboxy terminus.
  • Easily Screen and Profile CSK Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Content And Storage <-65°C
Quantity 10 μg