Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Promega DNA IQ™ Reference Sample Kit
Click to view available options
:
48 preparations
Description
- Optimal DNA Extraction from Forensic Casework and DNA Reference Samples
- Use DNA IQ™ Casework Pro Kit for: blood stains, semen stains, hairs, cigarette butts, tissue samples and trace or touch DNA
- Use DNA IQ™ Reference Sample Kit with: buccal swabs, FTA(R) blood card punches, liquid blood or other high-concentration samples
- Minimize need for concentration or dilution prior to amplification
Specifications
Specifications
Content And Storage | 15°C to 30°C |
For Use With (Equipment) | Maxwell™ 16 |
For Use With (Application) | Optimal DNA extraction from buccal swabs, FTA™ Blood card punches, Blood, DNA reference samples |
Includes | 48 DNA IQ™ reference sample cartridges, 50 plungers, 50 Elution tubes, 20mL Elution Buffer, 25mL Lysis Buffer, protocol |
No. of Reactions | 48 Preps |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction