Description
- Live-Cell Detection of Renilla Luminescence for up to 24 Hours
- Engineered coelenterazine for live-cell reporter gene, BRET and RNAi assays
- Automation-friendly and highly sensitive
- Patent pending.
- Certain applications of this product may require licenses from others.
- This product does not convey a license to use recombinant Renilla luciferase under U.S. Patents 5,292,658; 5,418,155 and related patents. Promega offers licensed Renilla luciferase vectors, which may be used in conjunction with this product.
Specifications
Specifications
| Content And Storage | -30°C to -10°C |
| Quantity | 0.34 mg |