Learn More
Promega EnduRen™ Live Cell Substrate
Catalog No. PRE6482 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
0.34 mg
3.4 mg
34 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PR-E6482 3.4 mg
PR-E6481 0.34 mg
PRE6485 34 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PR-E6482 Supplier Promega Supplier No. E6482
Only null left
Add to Cart
Add to Cart
An automation-friendly, highly sensitive substrate for Renilla luciferase-based gene reporter and BRET applications.
  • Live-Cell Detection of Renilla Luminescence for up to 24 Hours
  • Engineered coelenterazine for live-cell reporter gene, BRET and RNAi assays
  • Automation-friendly and highly sensitive
  • Patent pending.
  • Certain applications of this product may require licenses from others.
  • This product does not convey a license to use recombinant Renilla luciferase under U.S. Patents 5,292,658; 5,418,155 and related patents. Promega offers licensed Renilla luciferase vectors, which may be used in conjunction with this product.
Specifications
Content And Storage -30°C to -10°C
Quantity 3.4 mg