Learn More
Description
Includes
1 vial rLuciferase/Luciferin Reagent; 12mL L/L Reconstitution Buffer; 1 Protocol (TMF005)
- Detect and Quantitate ATP in Liquid Samples
†The method of recombinant expression of Coleoptera luciferase is covered by U.S. Patents 5,583,024; 5,674,713; and 5,700,673.
Specifications
Specifications
| Quantity | 100 Assays |
| Content And Storage | 1 Vial Luciferase/Luciferin reagent, 12mL L/L Reconstitution Buffer, 1 protocol. -30°C to -10°C |
| For Use With (Application) | For the rapid and quantitative detection of ATP in liquid samples |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.