Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Factor Xa Protease

Catalog No. PRV5581 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PR-V5581 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PR-V5581 Supplier Promega Supplier No. V5581
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Factor Xa Protease preferentially cleaves after the arginine residue in the amino acid sequence Ile-Glu-Gly-Arg.

  • Preferentially Cleaves After Arginine in the Amino Acid Sequence Ile-Glu-Gly-Arg
  • Purified from bovine plasma and activated with Russell's viper venom
  • Cleaves proteins and fusion proteins containing Factor Xa Protease sites

Specifications

Color Colorless
pH 7.4
For Use With (Application) Cleavage of Proteins and fusion Proteins containing Factor Xa Protease sites, including fusion Proteins generated with the PinPoint™ vectors
Storage Buffer 20mM Tris HCl (pH 7.4), 0.1M NaCl Reaction Buffer
Content And Storage -30°C to -10°C
Source Bovine Plasma
Quantity 50 μg
Product Type Factor Xa Protease
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.