Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega FGFR1 Kinase Enzyme System

Catalog No. PRV2991 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1/Ea.
10 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRV2991 10 μg
PRV9321 1/Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRV2991 Supplier Promega Supplier No. V2991
Add to Cart
Add to Cart

Recombinant human FGFR1 (amino acids 399-822) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. FGFR1 (also known as FLT2) is a member of the Fibroblast Growth Factor Receptor family.

  • Easily Screen and Profile FGFR1 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity

Specifications

Content And Storage <-65°C
For Use With (Application) Involved in mesoderm induction and patterning, cell growth and migration, organ formation and bone growth
Quantity 10 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.