Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega FluoroTect™ GreenLys In Vitro Translation Labeling System

Catalog No. PRL5001 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
40 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PR-L50-01 40 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PR-L50-01 Supplier Promega Supplier No. L5001
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

FluoroTect GreenLys tRNA, 40μL

FluoroTect Green(Lys) in vitro Translation Labeling System allows fluorescent labeling and detection of proteins synthesized in vitro. It is based on a lysine-charged tRNA labeled at the epsilon position of the lysine with the fluorophore BODIPY-FL.

  • Fluorescent Labeling and Detection of in Vitro Synthesized Proteins
  • Data in minutes versus overnight exposures associated with radioactivity-based systems
  • In-gel detection; No need to transfer, fix or dry gels
  • No radioactivity required

FluoroTect GreenLys incorporates dye conjugates made with the BODIPY-FL fluorescent reactive dyes, which are licensed from Molecular Probes, Inc., under U.S. Patent 5,433,896 for IVE analysis for research use only including GPR and ASR applications and Fluorotag™ technology and under U.S. Patent 6,306,628.

Specifications

Formulation 40μL FluoroTect greenLys tRNA
For Use With (Application) Fluorescent labeling and detection of Proteins synthesized in vitro
Includes 40μL FluoroTect greenLys tRNA
Content And Storage <−65°C
Product Type Labeling system
Quantity 40 Reactions
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.