Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega FYN A Kinase Enzyme System

Catalog No. PRV3571 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1/Ea.
10 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRV3571 10 μg
PRV9341 1/Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRV3571 Supplier Promega Supplier No. V3571
Add to Cart
Add to Cart

Includes

FYN A Kinase Enzyme System (V3571)

  • Kinase: FYN A, 10μg (Human, recombinant full-length). MW: ∽85kDa
  • Substrate: Poly (4:1 Glu, Tyr) Peptide.
  • Reaction Buffer, DTT, MnCl2
ADP-Glo™ Kinase Assay and EGFR Kinase Enzyme System (V9341)
  • 1 x 10μg EGFR Kinase Enzyme System (V3571) and 1 x 1000 ADP-Glo™ Kinase Assays (V9101)

Full-length recombinant human FYN A was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. FYN A (Fyn isoform A) is a member of the SRC tyrosine kinase oncogene family showing high homology to YES1, FGR and SRC.

  • Easily Screen and Profile FYN A Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity

Specifications

Content And Storage <-65°C
For Use With (Application) FYN by interaction with DAB1 regulates the Phosphorylation, trafficking and processing of APP and apoEr2
Quantity 10 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.