Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Promega GloMax™-Multi Jr Single-Tube Multimode Reader: Optical Kits
Click to view available options
Color:
Blue
GFPUV
Green
Red
UV
Description
- For Use With the GloMax(R)-Multi Jr Single-Tube Multimode Reader
- Instrument is discontinued
- Accessories (pending availability) and Service Agreement available until November 1, 2020
Specifications
Specifications
Color | UV |
For Use With (Equipment) | Single-Tube Multimode Reader |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction