Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega GoTaq™ Hot Start Polymerase: 5X

Catalog No. PRM5005 Shop All Promega Corporation Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
100U
2500U
500U
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRM5005 500U
PRM5001 100U
PRM5006 2500U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PRM5005 Supplier Promega Supplier No. M5005
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Includes

GoTaq Hot Start Polymerase is supplied with 5X Colorless GoTaq Flexi Buffer, 5X Green GoTaq Flexi Buffer, and 25mM MgCl2.

GoTaq™ Hot Start Polymerase contains the high-performance GoTaq™ DNA Polymerase bound to a proprietary antibody that blocks polymerase activity until the reaction is heated at 94-95°C for two minutes.

  • Robust PCR with Convenient Room-Temperature Setup
  • Minimize nonspecific amplification and primer-dimers
  • Optimize reaction conditions with supplied Flexi Reaction Buffer and MgCl2 solution
  • Direct-to-gel convenience
  • Supplied at a concentration of 5u/μL

  • U.S. Patent No. 6,242,235, Australian Patent No. 761757, Canadian Patent No. 2,335,153.
  • Licensed under U.S. Patent Nos. 5,338,671 and 5,587,287 and corresponding patents in other countries.

Specifications

Concentration 5 U/μL
Content And Storage 5X Hot start Polymerase (1 x 500U), colorless flexi Buffer (4 x 1mL), 5X green flexi Buffer (4x 1mL), MgCl2 (3 x 1.2mL). Store at −30 to −10°C
Quantity 500U
For Use With (Application) RT-PCR
Activation Time 2 min.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.