Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega GoTaq™ MDx Hot Start Polymerase

Catalog No. PRD6008 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10000U
100U
2500U
500U
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRD6008 10000U
PRD6001 100U
PRD6005 500U
PRD6006 2500U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. PRD6008 Supplier Promega Supplier No. D6008
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

GoTaq™ MDx Hot Start Polymerase contains GoTaq™ MDx DNA Polymerase bound to a proprietary antibody that blocks polymerase activity.

  • Inhibits Polymerase Activity at Temperatures below 70°C
  • Manufactured under cGMP
  • High-concentration and glycerol-free formulations also available
  • All formulations supplied at ≥5u/μL, except for high-concentration formulation supplied at ≥50u/μL
  • Custom formats and formulations

  • U.S. Pat. No. 6,242,235, Australian Pat. No. 761757, Canadian Pat. No. 2,335,153.
  • Licensed under U.S. Pat. Nos. 5,338,671 and 5,587,287 and corresponding patents in other countries.

Specifications

Content And Storage MgCl2 (60 x 750μL), MDx hot start Polymerase (20 x 500U), 5X colorless flexi Buffer (80 x 1mL), 5X green flexi Buffer (80 x 1mL). Store at −30 to −10°C
Quantity 10000U
For Use With (Application) Cloning
Activation Time 2 min.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.