Learn More
Promega GSK3β Kinase Enzyme System
Catalog No. PRV1991 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1/Ea.
10 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRV1991 10 μg
PRV9371 1/Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRV1991 Supplier Promega Supplier No. V1991
Only null left
Add to Cart
Add to Cart
Includes
GSK3β Kinase Enzyme System (V1991)
  • Kinase: GSK3β, 10μg (Human, recombinant full length). MW: ∽73kDa
  • Substrate: GSK3 derived from human muscle glycogen synthase 1 (amino acid 636–661)
  • Others: Reaction Buffer, DTT, MnCl
ADP-Glo™ Kinase Assay and EGFR Kinase Enzyme System (V9371)
  • 1 x 10μg GSK3β Kinase Enzyme System (V1991) and 1 x 1000 ADP-Glo™ Kinase Assays (V9101)
Recombinant full-length human GSK3 beta was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. GSK3 beta is a serine-threonine protein kinase originally identified as the kinase that phosphorylates and inhibits glycogen synthase.
  • Easily Screen and Profile GSK3β Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Content And Storage <-65°C
Quantity 10 μg