Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega GSK3α Kinase Enzyme System

Catalog No. PRV3051 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1/Ea.
10 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRV3051 10 μg
PRV9361 1/Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRV3051 Supplier Promega Supplier No. V3051
Add to Cart
Add to Cart

Includes

  • 1 x 10μg GSK3α Kinase Enzyme System (V3051) and 1 x 1000 ADP-Glo™ Kinase Assays (V9101)

Recombinant full-length human GSK3 alpha was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. GSK3 alpha is a multifunctional protein serine kinase implicated in the control of several regulatory proteins.

  • Easily Screen and Profile GSK3α Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity

Specifications

Molecular Weight (g/mol) 81
Content And Storage <-65°C
Quantity 10 μg
Form Fluid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.