Learn More
Description
- Covalent, Oriented Attachment of HaloTag(R) Fusion Proteins to a Solid Surface
- Covalent binding enables stringent washing, minimizes nonspecific background
- Fast binding kinetics enhance detection of protein:protein interactions, enable binding at low protein concentrations
- One mL of resin binds >7mg of HaloTag(R) fusion protein
Specifications
Specifications
| Type | HaloLink Resin |
| Sufficient For | Covalent and oriented attachment of HaloTag™ fusion Proteins to a solid surface |
| Quantity | 10 mL |
| Content And Storage | 2°C to 10°C |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.