Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega HaloTag™ Alexa Fluor™ 660 Fluorescent Ligand

Catalog No. PRG8472 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
15 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRG8472 15 μL
PRG8471 30 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRG8472 Supplier Promega Supplier No. G8472
Add to Cart
Add to Cart

The HaloTag™ Technology enables covalent protein labeling and immobilization in vivo and in vitro.

  • Covalently Attach Fluorescent Labels or Affinity Tags to Membrane or Intracellular Proteins
  • Cell permeant TMR, Oregon Green(R), diAcFAM and Coumarin ligands readily cross the cell membrane for labeling intracellular proteins
  • Cell impermeant tags include Alexa Fluor(R) 488 and 660 for quick cell surface labeling

Specifications

Concentration 3.5 mM
For Use With (Application) Protein localization, Protein trafficking, Protein capture, cell-to -gel analysis, Protein interaction analysis
Quantity 15 μL
Product Type HaloTag™ Fluorescent Ligand
Sufficient For For cell-surface labeling
Content And Storage -30°C to -10°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.