Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega HaloTag™ Amine (O4) Ligand

Catalog No. PRP6741
Click to view available options
Quantity:
5 mg

HaloTag™ Ligand Building Blocks are designed for use with HaloTag™ fusion proteins and can carry a variety of functionalities, including fluorescent labels, affinity tags and attachments to a solid phase.

  • Enables any Reporter, Protein or Nucleic Acid with Specified Functional Groups to be used with the HaloTag(R) Protein
  • Image live or fixed mammalian cells expressing HaloTag(R) protein or protein fusions
  • Analyze post-translational modification
  • Isolate proteins and protein complexes
  • Cat.# P6761 is discontinued

Specifications

Content And Storage -30°C to -10°C
For Use With (Equipment) Any reporter group, protein, or nucleic acid derivative containing an activated carboxylic acid, sulfonyl halide, or isocyanate
For Use With (Application) Any reporter group, Protein or Nucleic Acid derivative containing an activated Carboxylic Acid, Sulfonyl Halide or Isocyanate
Includes Fluorescent labels, affinity tags and attachments to a solid phase
Product Type Ligand Building Block
Quantity 5 mg
Form Solid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.