Learn More
Promega HaloTag™ Fusion (N-Terminal) Mammalian Expression Vectors
Shop All Promega Corporation Products
Description
- Append N-Terminal Tag for High-Level Protein Expression in Mammalian Cells
- Enables constitutive protein expression in mammalian cells
- Includes CMV immediate-early enhancer/promoter
- Suited to stable or transient gene expression
Specifications
Specifications
| Promoter | CMV |
| Protein Expression Level | High |
| Storage Buffer | 10mM Tris HCl (pH 7.4), 1mM EDTA |
| Content And Storage | -30°C to -10°C |
| Concentration | 1 μg/μL |
| Quantity | 20 μg |
| Vector | pHTN HaloTag CMV-neo |
| For Use With (Application) | Transient and stable gene expression, cell imaging of Protein localization or trafficking in conjunction |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.