Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega HaloTag™ Interchangeable Labeling Technology

Catalog No. PRG8592 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
15 μL
30 μL
Product Type:
HaloTag Alexa Fluor 488 Ligand
HaloTag Biotin Ligand
HaloTag Coumarin Ligand
HaloTag Oregon Green Ligand
HaloTag PEG-Biotin Ligand
HaloTag R110Direct Ligand
HaloTag TMR Ligand
HaloTag TMRDirect Ligand
HaloTag diAcFAM Ligand
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type
PRG8592 15 μL HaloTag PEG-Biotin Ligand
PRG1002 15 μL HaloTag Alexa Fluor 488 Ligand
PRG2802 15 μL HaloTag Oregon Green Ligand
PRG3221 30 μL HaloTag R110Direct Ligand
PRG2991 30 μL HaloTag TMRDirect Ligand
PRG8273 15 μL HaloTag diAcFAM Ligand
PRG8252 15 μL HaloTag TMR Ligand
PRG8282 15 μL HaloTag Biotin Ligand
PRG8582 15 μL HaloTag Coumarin Ligand
PRG1001 30 μL HaloTag Alexa Fluor 488 Ligand
PRG2801 30 μL HaloTag Oregon Green Ligand
PRG8251 30 μL HaloTag TMR Ligand
PRG8272 30 μL HaloTag diAcFAM Ligand
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Catalog No. PRG8592 Supplier Promega Supplier No. G8592
Add to Cart
Add to Cart

The HaloTag™ Technology enables covalent protein labeling and immobilization in vivo and in vitro.

  • Covalently Attach Fluorescent Labels or Affinity Tags to Membrane or Intracellular Proteins
  • Cell permeant TMR, Oregon Green(R), diAcFAM and Coumarin ligands readily cross the cell membrane for labeling intracellular proteins
  • Cell impermeant tags include Alexa Fluor(R) 488 and 660 for quick cell surface labeling

Specifications

Content And Storage -30°C to -10°C
For Use With (Application) Protein localization, Protein trafficking, Protein capture, Cell to Gel analysis, Protein to Protein interactions
Includes Fluorescent labels, affinity tags and attachments to a solid phase
Product Type HaloTag PEG-Biotin Ligand
Quantity 15 μL
Form Fluid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.