Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega HB101 Competent Cells

Catalog No. p-4907577
Click to view available options
Quantity:
20 x 50 μL
5 x 200 μL

HB101 cells are recA- to minimize recombination events and contain a restriction-minus background (hsdS20) to allow better representation when cloning methylated DNA. Suitable for vectors that do not require alpha-complementation for blue/white screening.

  • Ready-to-Use Competent E. coli Cells for Recombinant Cloning and Subcloning
  • Cells are ready to use—no preparation time necessary
  • Transformation efficiencies guaranteed
  • No worries about undesirable recombination events (recA- mutation)

Specifications

Concentration >108cfu/μg
Content And Storage Store at -70°C.
For Use With (Application) Transformation of unmethylated DNA derived from PCR, cDNA and many other sources
Includes HB101 Competent Cells (high efficiency) and Control DNA
pH 7.4
Product Type Competent Cells
Quantity 5 x 200 μL
Form Fluid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.