Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega HDAC-Glo™ I/II Assay and Screening Systems

Catalog No. PRG6422 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 mL
50 mL
100 mL
Product Type:
Assay
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type
PRG6422 100 mL Assay
PRG6420 10 mL Assay
PRG6421 50 mL Assay
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PRG6422 Supplier Promega Supplier No. G6422
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

The HDAC-Glo™ I/II Assays and Screening System are single-reagent-addition, homogeneous, luminescent assays that measure the relative activity of histone deacetylase (HDAC) class I and II enzymes from cells, extracts or purified enzyme sources.

  • Detects HDAC Class I and II Enzyme Activity
  • Simple add-mix-measure protocol
  • Use purified enzymes, extracts or cells directly in culture plates
  • Screening System includes HeLa Nuclear Extract

Specifications

Quantity 100 mL
pH 8
Product Type Assay
Content And Storage -30°C to -10°C
For Use With (Application) Determining HDAC inhibitor potency using purified enzymes, extracts or cells directly in culture plates
Form Fluid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.