Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Heat Block Adapter

Catalog No. PRA2661 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
No. per Pack:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. per Pack
PRA2661 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRA2661 Supplier Promega Supplier No. A2661
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Designed for a 2.2mL deep-well 96-well plate (Cat. # V6781) when using the Maxwell HT 96 gDNA Blood Isolation System (Cat.# A2670, A2671). Available for purchase as a standalone item.

  • Adapter For Use With 2.2mL Deep-Well 96-Well Plate
  • Used with the Maxwell(R) HT 96 gDNA Blood Isolation System (Cat.# A2670, A2671). Available for purchase as a standalone item.

Specifications

For Use With (Application) Specially designed for efficient heat transfer, interfaces with sbs format heaters
No. per Pack 1 Ea.
Storage Requirements Store at 15°C to 30°C
Product Type Heat Block Adapter
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.