Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega HER4 Kinase Enzyme System

Catalog No. PRV3101 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1/Ea.
10 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRV3101 10 μg
PRV9391 1/Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRV3101 Supplier Promega Supplier No. V3101
Add to Cart
Add to Cart

Includes

HER4 Kinase Enzyme System (V3101)

  • Kinase: HER4, 10μg (Human, recombinant; amino acids 682–993). MW: ∽57kDa
  • Substrate: Poly (4:1 Glu, Tyr) Peptide.
  • Others: Reaction Buffer, DTT, MnCl2
ADP-Glo™ Kinase Assay and HER4 Kinase Enzyme System (V9391)
  • 1 x 10μg EGFR Kinase Enzyme System (V3101) and 1 x 1000 ADP-Glo™ Kinase Assays (V9101)

Recombinant human HER4 (amino acids 682-993) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. HER4 or ERBB4 is a transmembrane receptor tyrosine kinase that belongs to the epidermal growth factor receptor family.

  • Easily Screen and Profile HER4 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity

Specifications

Content And Storage <-65°C
For Use With (Application) Interaction with its activating ligand, heregulin, HER4 can regulate proliferation and differentiation of cells
Quantity 10 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.