Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega IGF1R Kinase Enzyme System

Catalog No. PRV3581 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1/Ea.
10 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRV3581 10 μg
PRV9401 1/Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRV3581 Supplier Promega Supplier No. V3581
Add to Cart
Add to Cart

Includes

IGF1R Kinase Enzyme System (V3581)

  • Kinase: IGF1R, 10μg (Human, recombinant; amino acids 960–end). MW: ∽53kDa
  • Substrate: IGF1Rtide (KKKSPGEYVNIEFG); derived from human IRS-1 protein residues 891–902
  • Reaction Buffer, DTT, MnCl2
ADP-Glo™ Kinase Assay and EGFR Kinase Enzyme System (V9401)
  • 1 x 10μg EGFR Kinase Enzyme System (V3581) and 1 x 1000 ADP-Glo™ Kinase Assays (V9101)

Recombinant human IGF1R was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag. IGF1R (Insulin-like Growth Factor 1) is a transmembrane tyrosine kinase receptor that is activated by IGF-1 and by the related growth factor IGF-2.

  • Easily Screen and Profile IGF1R Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity

Specifications

Molecular Weight (g/mol) 53
Content And Storage <-65°C
For Use With (Application) Growth and anabolic effects in adults
Quantity 10 μg
Form Fluid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.