Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega IGF1R Kinase Enzyme System

Catalog No. p-4671673
Click to view available options
Quantity:
1/Ea.
10 μg

Includes

IGF1R Kinase Enzyme System (V3581)

  • Kinase: IGF1R, 10μg (Human, recombinant; amino acids 960–end). MW: ∽53kDa
  • Substrate: IGF1Rtide (KKKSPGEYVNIEFG); derived from human IRS-1 protein residues 891–902
  • Reaction Buffer, DTT, MnCl2
ADP-Glo™ Kinase Assay and EGFR Kinase Enzyme System (V9401)
  • 1 x 10μg EGFR Kinase Enzyme System (V3581) and 1 x 1000 ADP-Glo™ Kinase Assays (V9101)

Recombinant human IGF1R was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag. IGF1R (Insulin-like Growth Factor 1) is a transmembrane tyrosine kinase receptor that is activated by IGF-1 and by the related growth factor IGF-2.

  • Easily Screen and Profile IGF1R Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity

Specifications

Molecular Weight (g/mol) 53
Content And Storage <-65°C
For Use With (Application) Growth and anabolic effects in adults
Quantity 10 μg
Form Fluid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.